Skip to Content
Merck
All Photos(2)

Documents

07-850

Sigma-Aldrich

Anti-phospho-LIMK 1/2 (Tyr507/Thr508) Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr507/pThr508)

Gene Information

human ... LIMK1(3984)

Specificity

Predicted to cross-react with Mouse, Rat, and Frog (100% homologous) based on sequence homology.
Recognizes phosphorylated LIMK1/2 (Tyr507/Thr508)

Immunogen

Peptide corresponding to amino acid region encompassing the human, mouse, rat and chicken phospho-LIMK1/2 (Tyr507/Thr508)

Application

Detect phospho-LIMK 1/2 (Tyr507/Thr508) using this anti-phospho-LIMK 1/2 (Tyr507/Thr508) Antibody validated for use in WB.
Immunoblot Analysis: A 1:1000 dilution of this lot detected phosphorylated LIMK 1/2 (Tyr507/Thr508) in RIPA lysates of SDF-1α treated Jurkat cells.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

Routinely evaluated by immunoblot.

Target description

72 kDa (LIMK1) and 65 kDa (LIMK2)

Physical form

100 μL of affinity purified rabbit polyclonal IgG in 50% storage buffer (PBS (without Mg2+ and Ca2+), pH 7.3 containing 1.0 mg/mL BSA (IgG, protease free) and 0.05% sodium azide) and 50% glycerol.
ImmunoAffinity Purified

Storage and Stability

Stable for 2 years at -20°C from date of shipment. Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol-containing solutions to become frozen during storage.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliana Soosairajah et al.
The EMBO journal, 24(3), 473-486 (2005-01-22)
Slingshot (SSH) phosphatases and LIM kinases (LIMK) regulate actin dynamics via a reversible phosphorylation (inactivation) of serine 3 in actin-depolymerizing factor (ADF) and cofilin. Here we demonstrate that a multi-protein complex consisting of SSH-1L, LIMK1, actin, and the scaffolding protein
Hirobumi Tada et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(45), E7097-E7105 (2016-10-30)
Social separation early in life can lead to the development of impaired interpersonal relationships and profound social disorders. However, the underlying cellular and molecular mechanisms involved are largely unknown. Here, we found that isolation of neonatal rats induced glucocorticoid-dependent social
Down-regulation of PKCzeta expression inhibits chemotaxis signal transduction in human lung cancer cells.
Ying Liu,Bing Wang,Jingna Wang,Wuzhou Wan,Ronghua Sun,Yuliang Zhao,Ning Zhang
Lung Cancer null
D C Edwards et al.
Nature cell biology, 1(5), 253-259 (1999-11-13)
Extracellular signals regulate actin dynamics through small GTPases of the Rho/Rac/Cdc42 (p21) family. Here we show that p21-activated kinase (Pak1) phosphorylates LIM-kinase at threonine residue 508 within LIM-kinase's activation loop, and increases LIM-kinase-mediated phosphorylation of the actin-regulatory protein cofilin tenfold
K Ohashi et al.
The Journal of biological chemistry, 275(5), 3577-3582 (2000-02-01)
LIM-kinase 1 (LIMK1) phosphorylates cofilin, an actin-depolymerizing factor, and regulates actin cytoskeletal reorganization. LIMK1 is activated by the small GTPase Rho and its downstream protein kinase ROCK. We now report the site of phosphorylation of LIMK1 by ROCK. In vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service