Skip to Content
Merck
All Photos(1)

Key Documents

07-639

Sigma-Aldrich

Anti-PRMT7 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Prmt7(214572)

Specificity

Also known to cross-react with chimpanzee.
PRMT7

Immunogen

KLH-conjugated, synthetic peptide (KIFCSRANPTTGSVEWLEEDEHYD) corresponding to amino acids 2-25 of human Prmt7.

Application

Anti-PRMT7 Antibody is a high quality Rabbit Polyclonal Antibody for the detection of PRMT7 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated by immunoblot on RIPA lysates from HEK293 cells.

Target description

85kDa

Physical form

70% storage buffer (0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide) and 30% glycerol
Format: Purified
Protein A purified

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of new drug sensitivity genes using genetic suppressor elements: protein arginine N-methyltransferase mediates cell sensitivity to DNA-damaging agents.
Gros, Laurent, et al.
Cancer Research, 63, 164-171 (2003)
Sung-Hun Lee et al.
Nucleic acids research, 44(22), 10603-10618 (2016-09-15)
Self-renewal and pluripotency are two fundamental characteristics of embryonic stem cells (ESCs) and are controlled by diverse regulatory factors, including pluripotent factors, epigenetic regulators and microRNAs (miRNAs). Although histone methyltransferases are key epigenetic regulators, whether and how a histone methyltransferase
Human protein arginine methyltransferases in vivo--distinct properties of eight canonical members of the PRMT family.
Herrmann, F; Pably, P; Eckerich, C; Bedford, MT; Fackelmayer, FO
Journal of Cell Science null
Christina Demetriadou et al.
Cell death & disease, 10(3), 236-236 (2019-03-13)
N-alpha-acetyltransferase 40 (NAA40) catalyzes the transfer of an acetyl moiety to the alpha-amino group of serine 1 (S1) on histones H4 and H2A. Our previous studies linked NAA40 and its corresponding N-terminal acetylation of histone H4 (N-acH4) to colorectal cancer
Tina Branscombe Miranda et al.
The Journal of biological chemistry, 279(22), 22902-22907 (2004-03-27)
We have identified a mammalian arginine N-methyltransferase, PRMT7, that can catalyze the formation of omega-NG-monomethylarginine in peptides. This protein is encoded by a gene on human chromosome 16q22.1 (human locus AK001502). We expressed a full-length human cDNA construct in Escherichia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service