Skip to Content
Merck

Skip To

07-609-I

Anti-Peroxiredoxin 1

from rabbit, purified by affinity chromatography

Synonym(s):

EC:1.11.1.15, Natural killer cell-enhancing factor A, NKEF-A, Proliferation-associated gene protein, PAG, Thioredoxin peroxidase 2, Thioredoxin-dependent peroxide reductase 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PRDX1(5052)

General description

Peroxiredoxin-1 (UniProt: Q06830; also known as EC:1.11.1.15, Natural killer cell-enhancing factor A, NKEF-A, Proliferation-associated gene protein, PAG, Thioredoxin peroxidase 2, Thioredoxin-dependent peroxide reductase 2) is encoded by the PRDX1 (also known as PAGA, PAGB, TDPX2) gene (Gene ID: 5052 ) in human. Peroxiredoxins belong to a family of non-seleno peroxidases. Thus far six different mammalian isoforms have been reported and they all catalyze peroxide reduction of hydrogen peroxide, organic hydroperoxides and peroxynitrite. All peroxiredoxins share a conserved Cys residue that corresponds to the N-terminal Cys51 in mammalian peroxiredoxin-1. Peroxiredoxin 1-4 also contain an additional conserved Cys residue in the C-terminal region, Cys172. The N-terminal Cys51 is shown to be oxidized by hydrogen peroxide to form cysteine-sulfenic acid (Cys51-SOH) and reacts with Cys172-SH of the other subunit to produce an intermolecular disulfide, which can only be reduced by thioredoxin and not by glutathione or glutaredoxin. Peroxiredoxin-1 is constitutively expressed in most human cells and is induced to higher levels upon serum stimulation in cells. It is involved in redox regulation of the cell and important role in overcoming oxidative stress. Peroxiredoxin-1 also eliminates peroxides generated during drug metabolism. It is phosphorylated on Thr90 during the M-phase of cell cycle, which leads to over 80% reduction in its enzymatic activity.
~21 kDa observed; 22.11 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

KLH-conjugated linnear peptide corresponding to 12 amino acids from the C-terminal region of human Peroxiredoxin-1.

Application

Anti-Peroxiredoxin 1 Antibody, Cat. No. 07-609-I, is a highly specific rabbit polyclonal antibody that targets Peroxiredoxin 1 and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin) and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Peroxiredoxin 1 in HeLa cells.

Immunohistochemistry Analysis: A 1:250-1:1,000 dilution from a representative lot detected Peroxiredoxin 1 in human kidney, human small intestine, and human cerebellum tissues.
Research Category
Neuroscience

Biochem/physiol Actions

Predicted to react with various other mammalian species, including Bat, Canine, Hamster, Rat, and Rhesus Monkey.
This polyclonal antibody detects Peroxiredoxin-1 by Western blotting and immunocytochemistry in in DU145 cells and HeLa cells, respectively.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in DU145 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Peroxiredoxin 1 in 10 µg of DU145 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.
Replaces: 07-609

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters