Skip to Content
Merck
All Photos(2)

Key Documents

07-550

Sigma-Aldrich

Anti-SUV39H1 Antibody

Upstate®, from rabbit

Synonym(s):

KMT1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SUV39H1(6839)

Specificity

SUV39H1

Immunogen

Peptide (C-QVDPVDMESTRMDSNFGLAGLPGSP) corresponding to amino acids 368-392 of mouse suppressor of variegation 3-9 homolog 1 (SUV39H1).

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-SUV39H1 Antibody (Rabbit Polyclonal Antibody) validated in WB to detect SUV39H1 also known as KMT1A, Histone H3-K9 Methyltransferase 1.

Quality

Routinely evaluated by immunoblot in U20S cells lysed in RIPA with 0.1% SDS and sonicated.

Target description

48kDa

Linkage

Replaces: MABE552

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, pH 7.4.
Format: Purified
Protein A purified

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hakima Yahi et al.
The Journal of biological chemistry, 283(35), 23692-23700 (2008-07-05)
Mechanisms of transcriptional repression are important during cell differentiation. Mammalian heterochromatin protein 1 isoforms HP1alpha, HP1beta, and HP1gamma play important roles in the regulation of chromatin structure and function. We explored the possibility of different roles for the three HP1
Imtiyaz Yaseen et al.
The EMBO journal, 37(2), 183-200 (2017-11-25)
Host cell defense against an invading pathogen depends upon various multifactorial mechanisms, several of which remain undiscovered. Here, we report a novel defense mechanism against mycobacterial infection that utilizes the histone methyltransferase, SUV39H1. Normally, a part of the host chromatin
Rajeev Mishra et al.
3 Biotech, 12(11), 290-290 (2022-10-25)
Telomere length is primarily controlled by the enzyme telomerase, but being chromatin structures, telomeres undergo epigenetic regulation for their maintenance and function. Altered telomere length among cancer cells combined with shorter telomere length in cancer-associated stromal cells, strongly implicated with
S J Nielsen et al.
Nature, 412(6846), 561-565 (2001-08-03)
In eukaryotic cells the histone methylase SUV39H1 and the methyl-lysine binding protein HP1 functionally interact to repress transcription at heterochromatic sites. Lysine 9 of histone H3 is methylated by SUV39H1 (ref. 2), creating a binding site for the chromo domain
R Firestein et al.
Molecular and cellular biology, 20(13), 4900-4909 (2000-06-10)
Mammalian SET domain-containing proteins define a distinctive class of chromatin-associated factors that are targets for growth control signals and oncogenic activation. SUV39H1, a mammalian ortholog of Drosophila Su(var)3-9, contains both SET and chromo domains, signature motifs for proteins that contribute

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service