Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse, human
manufacturer/tradename
Upstate®
technique(s)
immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... ELAVL1(1994)
General description
Immunogen
Application
10 µg from a representative lot detected HuR in RIPA lysate.
Immunocytochemistry Analysis:
A 1:500 dilution from a representative lot detected HuR in HeLa and NIH/3T3 cell lysates.
Immunohistochemistry Analysis:
This antibody has been reported by an independent laboratory to detect HuR in rabbit placental tissue.
Epigenetics & Nuclear Function
Transcription Factors
RNA Binding Protein (RBP)
Biochem/physiol Actions
Physical form
Preparation Note
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.
Analysis Note
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| antibody form purified antibody | antibody form affinity isolated antibody | antibody form affinity purified immunoglobulin | antibody form affinity purified immunoglobulin |
| species reactivity mouse, human | species reactivity human, rat, mouse | species reactivity mouse, human | species reactivity mouse, human |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
Still not finding the right product?
Explore all of our products under Anti-HuR Antibody
— or —
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



