Skip to Content
Merck
All Photos(1)

Key Documents

07-394

Sigma-Aldrich

Anti-MAGE-D1/NRAGE Antibody

serum, Upstate®

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€547.00

€547.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
€547.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€547.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MAGED1(9500)

Specificity

NRAGE/MAGE-D1

Immunogen

GST fusion protein corresponding to residues 53-295 of rat MAGE-D1/NRAGE

Application

Anti-MAGE-D1/NRAGE Antibody is an antibody against MAGE-D1/NRAGE for use in IP, WB, IH.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Developmental Neuroscience

Quality

routinely evaluated by immunoblot in RIPA lysates from PC12 cells

Target description

90-96 kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, before the addition of glycerol to 30%
Antiserum

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amir H Salehi et al.
The Journal of biological chemistry, 277(50), 48043-48050 (2002-10-12)
The p75 neurotrophin receptor (p75NTR) mediates signaling events leading to activation of the JNK pathway and cell death in a variety of cell types. We recently identified NRAGE, a protein that directly interacts with the p75NTR cytosolic region and facilitates
A H Salehi et al.
Neuron, 27(2), 279-288 (2000-09-14)
The mechanisms employed by the p75 neurotrophin receptor (p75NTR) to mediate neurotrophin-dependent apoptosis are poorly defined. Two-hybrid analyses were used to identify proteins involved in p75NTR apoptotic signaling, and a p75NTR binding partner termed NRAGE (for neurotrophin receptor-interacting MAGE homolog)
Jennifer M Doyle et al.
Molecular cell, 39(6), 963-974 (2010-09-25)
The melanoma antigen (MAGE) family consists of more than 60 genes, many of which are cancer-testis antigens that are highly expressed in cancer and play a critical role in tumorigenesis. However, the biochemical and cellular functions of this enigmatic family

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service