Skip to Content
Merck
All Photos(1)

Key Documents

07-374

Sigma-Aldrich

Anti-phospho-KDR/Flk-1/VEGFR2 (Tyr1214) Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€347.00

€347.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
€347.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€347.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr1214)

Gene Information

human ... KDR(3791)

Specificity

phospho-KDR/VEGFR2 (Tyr1214)

Immunogen

(...FHpYDN...) in which pY corresponds to phosphorylated tyrosine at residue 1214 of human KDR/VEGFR2. Tyr1214 of human KDR corresponds to Tyr1212 in the mouse protein.

Application

Anti-phospho-KDR/Flk-1/VEGFR2 (Tyr1214) Antibody detects level of phospho-KDR/Flk-1/VEGFR2 (Tyr1214) & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Quality

routinely evaluated by immunoblot RIPA lysates from porcine aortic endothelial cells expressing human CSF-1R/murineVEGFR-2 chimera

Target description

200-230 kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl,0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein A purified

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Substitution of C-terminus of VEGFR-2 with VEGFR-1 promotes VEGFR-1 activation and endothelial cell proliferation.
Meyer, Rosana D, et al.
Oncogene, 23, 5523-5531 (2004)
Michael J Donovan et al.
European journal of cancer (Oxford, England : 1990), 45(8), 1518-1526 (2009-03-11)
To identify clinical and biometric features associated with overall survival of patients with advanced refractory non-small-cell lung cancer (NSCLC) treated with gefitinib. One hundred and nine diagnostic NSCLC samples were analysed for EGFR mutation status, EGFR immunohistochemistry, histologic morphometry and
V Dayanir et al.
The Journal of biological chemistry, 276(21), 17686-17692 (2001-03-30)
Activation of vascular endothelial growth factor receptor-2 (VEGFR-2) plays a critical role in vasculogenesis and angiogenesis. However, the mechanism by which VEGFR-2 activation elicits these cellular events is not fully understood. We recently constructed a chimeric receptor containing the extracellular
N Rahimi et al.
The Journal of biological chemistry, 275(22), 16986-16992 (2000-04-05)
Vascular endothelial growth factor (VEGF) provokes angiogenesis in vivo and stimulates growth and differentiation of endothelial cells in vitro. Although VEGF receptor-1 (VEGFR-1) and VEGFR-2 are known to be high affinity receptors for VEGF, it is not clear which of
Rosana D Meyer et al.
The Journal of biological chemistry, 277(30), 27081-27087 (2002-05-25)
Vascular endothelial growth factor receptor (VEGFR)-2 plays a critical role in vasculogenesis during embryonic development and pathological angiogenesis, but little is known about the molecular mechanisms governing its functions. Here we investigated the role of tyrosine 1212 on mouse VEGFR-2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service