Sign In to View Organizational & Contract Pricing.
About This Item
eCl@ss:
32160702
UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-
biological source
rabbit
conjugate
unconjugated
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
mouse, human
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... OSR1(130497), OXSR1(9943)
General description
Odd-skipped-related protein 1 (OSR1) is expressed during the beginning stages of embryogenesis in the intermediate mesoderm and in patterns during limb and branchial arch development. It has been shown that OSR1 plays a critical part in creating kidney precursor cells and regulating differentiation into kidney tubular tissue. OSR1 may play a role in limiting endoderm differentiation from mesendoderm. Expression of OSR1 is controlled by Runx2 and Ikzf1 and is key in development of embryonic, heart and urogenital tissue.
~58 kDa observed
Immunogen
GST-tagged recombinant protein corresponding to human OSR1.
Application
Detect the OSR1 protein using this Anti-OSR1 validated for use in WB.
Research Category
Signaling
Signaling
Research Sub Category
Developmental Signaling
Developmental Signaling
Biochem/physiol Actions
Other homologies: Rat (96% sequence homology).
Physical form
Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Control
HEK293 cell lysate
HEK293 cell lysate
Evaluated by Western Blot in HEK293 cell lysate.
Western Blot Analysis: 1:1,000 dilution of this antibody detected OSR1 on 10 µg of HEK293 cell lysate.
Western Blot Analysis: 1:1,000 dilution of this antibody detected OSR1 on 10 µg of HEK293 cell lysate.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.