Skip to Content
Merck

Skip To

07-208

Anti-AIF (Apoptosis Inducing Factor) Antibody

Upstate®, from rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
3
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... AIFM1(9131)

General description

57kDa

Immunogen

peptide corresponding to amino acids 593-606 of human Apoptosis Inducing Factor, AIF (C-KDGEQHEDLNEVAK), containing a N-terminal cysteine

Application

Anti-AIF (Apoptosis Inducing Factor) Antibody detects level of AIF (Apoptosis Inducing Factor) & has been published & validated for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

AIF

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein A chromatography

Preparation Note

2 years at -20°C

Analysis Note

routinely evaluated by immunoblot on a mouse liver preparation

Other Notes

Replaces: 04-430

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB16502AB16501A7549
species reactivity

human, rat, mouse

species reactivity

human

species reactivity

human, mouse, rat

species reactivity

mouse, human

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

IgG fraction of antiserum

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

microarray: suitable, western blot: 1:1,000 using human epitheloid carcinoma HeLa cell extract and mouse brain extract


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters