Skip to Content
Merck
All Photos(1)

Key Documents

07-1799

Sigma-Aldrich

Anti-NCoA-2 Antibody

from rabbit

Synonym(s):

Transcriptional intermediary factor 2, glucocorticoid receptor-interacting protein-1, nuclear receptor coactivator 2, glucocorticoid receptor interacting protein 1, steroid receptor coactivator-2, transcriptional mediators/intermediary factor 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€421.00

€421.00


Estimated to ship on09 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on09 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NCOA2(10499)

General description

NCoA2 (nuclear receptor coactivator 2) is a transcriptional coactivator for steroid receptors and nuclear receptors. Also known as glucocorticoid receptor interacting protein 1 (GRIP1), steroid receptor coactivator-2 (SRC-2), or transcriptional mediators/intermediary factor 2 (TIF2), this coregulatory protein contains several nuclear receptor interacting domains and an intrinsic histone acetyltransferase activity. NCoA-2 assists nuclear receptors in the upregulation of DNA expression. In cells ligand activated nuclear receptors recruit NCoA-2 to DNA where it acylates histones to make targeted regions accessible to transcriptional machinery.

Specificity

Homology to mouse and rat is 95%.
This antibody recognizes NCoA-2.

Immunogen

Epitope: Unknown
Linear peptide corresponding to human NCoA-2

Application

Chromatin Immunoprecipitation:
A representative lot of this antibody was used for chIP. See Monroe, D.G., et al. (2006) J. Biol. Chem. 281(39):28596-28604.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-NCoA-2 Antibody (rabbit polyclonal antibody) validated in WB to detect NCoA-2 also known as Transcriptional intermediary factor 2, glucocorticoid receptor-interacting protein-1, nuclear receptor coactivator 2.

Quality

Evaluated by Western Blot in U2 OS cell lysate.

Western Blot Analysis: 0.5 µg/ml of this antibody detected SRC2 on 10 µg of U2 OS cell lysate.

Target description

~ 160 kDa

Physical form

Affinity purified
Format: Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
U2 OS cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service