Skip to Content
Merck
All Photos(1)

Key Documents

07-1708

Sigma-Aldrich

Anti-FOXH1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Forkhead activin signal transducer 1, Forkhead activin signal transducer 2, TGF-beta/activin signal transducer, forkhead activin signal transducer-1, forkhead box H1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€316.00

€316.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€316.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€316.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXH1(8928)
mouse ... Foxh1(14106)

General description

Forkhead box H1 (FOXH1) is a member of the HNF–3FKH family of transcriptional activators. FOXH1 is a winged helix with two loops on the C-terminal side expressed in all human tissue. FOXH1 is an embryonic transcription regulator and is an essential transcriptional co-activator that binds with SMAD2 to activate activin, and therefore induce the gooseciod (GSC) promoter. After FOXH1 binds to SMAD4, activin response element is activated and TGF-β is stimulated. FOXH1 also interacts with NKX2-5 to form an essential component of anterior heart field development. FOXH1 is associated with complex brain malfunctions due to incomplete cleavage of the prosencephalon, and in developmental delays resulting in facial abnormalities such as cyclopia and severe cleft lip or palate.

Specificity

This antibody recognizes FOXH1.

Immunogen

Synthetic to peptide corresponding to FOXH1.

Application

Anti-FOXH1 Antibody is a Rabbit Polyclonal Antibody for detection of FOXH1 also known as Forkhead activin signal transducer, TGF-beta/activin signal transduce & has been validated in ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by immunocytochemistry on NIH/3T3, HeLa, and A431 cells.

Immunocytochemistry:
1:500 dilution of this antibody was used to detect FOXH1 in NIH/3T3, HeLa and A431 cells.

Target description

38 kDa

Physical form

Affinity purified
Purified rabbit polyclonal antibody precipitated in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.
To reconstitute the antibody, centrifuge the antibody at a moderate speed (5,000 rpm) for 5 minutes. Carefully remove the ammonium sulfate/PBS buffer solution and discard; 10μL of residual ammonium sulfate solution will not affect the re-suspension of the antibody. Do not let the protein pellet dry, as severe loss of antibody reactivity can occur. Re-suspend the antibody pellet in 100L either standard PBS or TBS (pH 7.3-7.5). DO NOT VORTEX. Mix by gentle stirring with a wide pipet tip or gentle finger-tapping. Let the precipitated antibody rehydrate for 1 hour at 4-25°C prior to use. Small particles of precipitated antibody that fail to re-suspend are normal. Vials are overfilled to compensate for any losses.

Analysis Note

Control
NIH/3T3, HeLa, and A431 cells.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service