Skip to Content
Merck
All Photos(2)

Documents

07-1673

Sigma-Aldrich

Anti-SOX1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse

technique(s)

immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Sox-1 protein (SOX1), also known as SRY (Sex determining region Y)-box 1, is expressed in the developing brain. SOX1 is related to the HMG box of family transcription factors, which are a highly motile group, with a highly conserved DNA binding domain. Sox1 utilizes multiple pathways to play a direct role on cell fate. SOX1 is also directly involved with sex determination of the Y chromosome. Binding of the SOX1, C-terminus, to the Hes 1 promoter results in Hes1 suppression and attenuation of Notch signaling. When the C-terminus of SOX1 binds to β-catenin, TCF/LEF is suppressed, resulting in the attenuation of Wnt signaling.

Specificity

This antibody recognizes SOX1.

Immunogen

Synthetic peptide corresponding to SOX1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-SOX1 Antibody (Rabbit Polyclonal Antibody) validated in IHC(P) to detect SOX1 also known as SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1.

Quality

Evaluated by immunohistochemistry on human epidermis.

Immunohistochemistry(paraffin):
A 1:300 dilution of this antibody was used to detect SOX1 in human epidermis.

Physical form

Affinity purified
Purified rabbit polyclonal antibody precipitated in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.
To reconstitute the antibody, centrifuge the antibody at a moderate speed (5,000 rpm) for 5 minutes. Carefully remove the ammonium sulfate/PBS buffer solution and discard; 10 μL of residual ammonium sulfate solution will not affect the re-suspension of the antibody. Do not let the protein pellet dry, as severe loss of antibody reactivity can occur. Re-suspend the antibody pellet in 100L either standard PBS or TBS (pH 7.3-7.5). DO NOT VORTEX. Mix by gentle stirring with a wide pipet tip or gentle finger-tapping. Let the precipitated antibody rehydrate for 1 hour at 25°C prior to use. Small particles of precipitated antibody that fail to re-suspend are normal. Vials are overfilled to compensate for any losses.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Sook Shin et al.
Scientific reports, 12(1), 17605-17605 (2022-10-21)
Many cell-based therapies are challenged by the poor localization of introduced cells and the use of biomaterial scaffolds with questionable biocompatibility or bio-functionality. Endothelial progenitor cells (EPCs), a popular cell type used in cell-based therapies due to their robust angiogenic
Francesco Neri et al.
Molecular and cellular biology, 32(4), 840-851 (2011-12-21)
Myc family members are critical to maintain embryonic stem cells (ESC) in the undifferentiated state. However, the mechanism by which they perform this task has not yet been elucidated. Here we show that Myc directly upregulates the transcription of all

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service