Skip to Content
Merck

Skip To

07-167

Anti-Blys/TALL-1/BAFF/THANK Antibody

Upstate®, from rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
17
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TNFSF13B(10673)

General description

32kDa

Immunogen

peptide corresponding to amino acids 254-269 of human Blys (EEGDELQLAIPRENAQ)

Application

Anti-Blys/TALL-1/BAFF/THANK Antibody is an antibody against Blys/TALL-1/BAFF/THANK for use in WB.

Biochem/physiol Actions

Blys/TALL-1/BAFF/THANK

Physical form

Format: Purified
PBS containing 0.05% BSA and 0.05% sodium azide.

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.
routinely evaluated by immunoblot on RIPA lysates from Jurkat cells or HeLa nuclear extract

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters