Skip to Content
Merck
All Photos(3)

Key Documents

07-1500

Sigma-Aldrich

Anti-G Protein Giα 1/2 Antibody

from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€403.00

€403.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€403.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, bovine, mouse

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

bovine ... Gnai1(281790)
human ... GNAI1(2770)
mouse ... Gnai1(14677)
rat ... Gnai1(25686)

General description

GTP-binding proteins (G-proteins) play a critical role in signal transduction by coupling receptors to effectors. The intracellular consequences of G-protein activation include second messenger generation, protein phosphorylation, ion channel activation, gene induction, cell growth, and differentiation. Gi belongs to the pertussis toxin sensitive G-proteins. Gi consists of three different subtypes of alpha-subunits (alphai1, alphai2 and alphai3).

Specificity

Recognizes G protein subunit αi1.

Immunogen

Peptide corresponding to amino acids 345-354 of Gi1 and 346-355 of Gi2

Application

Anti-G Protein Giα 1/2 Antibody is an antibody against G Protein Giα 1/2 for use in WB, IH(P).
Immunohistochemistry(paraffin):
Representative testing from a previous lot.
Optimal Staining of GI ALPHA 1/2 With Citrate pH 6.0 Epitope Retrieval: Rat Cerebellum GI ALPHA 1/2 representative staining pattern/morphology in rat cerebellum. Tissue was pretreated with citrate pH 6, antigen retrieval. A previous lot of this antibody was diluted to 1:100, using IHC-Select Detection with HRP-DAB.

Optimal working dilutions must be determined by end user.

Quality

Routinely evaluated by Western Blot on L6 lysates.

Western Blot Analysis: 1:500 dilution of this lot detected G protein Gialpha 1/2 on 10 μg of L6 lysates.

Target description

~40 kDa

Linkage

Replaces: AB4067

Physical form

Format: Purified

Storage and Stability

Stable at -20°C in undiluted aliquots for up to 12 months from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service