Skip to Content
Merck
All Photos(4)

Key Documents

07-1223

Sigma-Aldrich

Anti-Protein Phosphatase 2 C α/β Antibody

from rabbit

Synonym(s):

Protein phosphatase 2C, Pyruvate dehydrogenase, Pyruvate dehydrogenase phosphatase, catalytic subunit 1, Lipoamide-phosphatase 1, mitochondrial precursor, Protein phosphatase 2C, magnesium-dependent, catalytic subunit, Pyruvate dehydrogenase (Lipoamide)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€220.00

€220.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€220.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€220.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

bovine, human, rat

species reactivity (predicted by homology)

mouse

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PDP1(54704)

General description

PP-2 C α/β is heterodimer located in the mitochondrial matrix. Within the pyruvate dehydrogenase complex, PP-2 C α/β serves to catalyze the dephosphorylation and concomitant reactivation of the E1 component’s alpha subunit. Mutations occurring in the PDP1 gene are the cause of a disease known as pyruvate dehydrogenase deficiency (PDP deficiency), which leads to neurological dysfunction from lactic acidosis.

Specificity

Recognizes the alpha/beta regulatory subunit isotype of protein phosphatase 2C (PP2C). PP 2C alpha/beta polyclonal antibody binds a 44-46 kDa protein.

Immunogen

Purified peptide conjugated to KLH.

Application

Detect Protein Phosphatase 2 C α/β using this Anti-Protein Phosphatase 2 C α/β Antibody validated for use in WB.
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases

Quality

Evaluated by Western Blot in L6 cell lysates.
Western Blot Analysis: 1:500 dilution of this lot detected PP-2 C ALPHA/BETA on 10 ug of L6 lysates.

Target description

Approx. 46 kDa

Linkage

Replaces: AB4090

Physical form

Format: Purified
Protein Purified
Purified immunoglobulin in phosphate buffered saline with 0.08% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
L6 Cell Lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service