Skip to Content
Merck
All Photos(3)

Key Documents

06-1331

Sigma-Aldrich

Anti-IL-23R Antibody, cytoplasmic

from rabbit, purified by affinity chromatography

Synonym(s):

interleukin 23 receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€408.00

€408.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IL23R(149233)

General description

Interleukin 23 receptor (IL-23R) is a type I cytokine receptor that is a subunit of the receptor for IL23A/IL23. IL-23R pairs with the receptor molecule IL12RB1/IL12Rbeta1, and both are required for IL23A signaling. IL-23R associates constitutively with Janus kinase 2 (JAK2), and also binds to transcription activator STAT3.

Specificity

This antibody recognizes the cytoplasmic domain of IL-23R.
May detect other isoforms of IL-23R (see molecular weight).

Immunogen

KLH-conjugated linear peptide corresponding to human IL-23R at and around the cytoplasmic domain.

Application

Anti-IL-23R Antibody, cytoplasmic detects level of IL-23R & has been published & validated for use in WB, IH, ELISA, FC.
Does not work for mouse in flow cytometry.
Immunohistochemistry Analysis: A 1:100 dilution from a previous lot detected IL-23R in mouse spleen tissue.

Flow Cytometry Analysis: 2-10 µg from a previous lot detected IL-23R in human PBMCs. Does not work in mouse for flow cytometry.

ELISA: A previous lot of this antibody was tested on ELISA by an independent laboratory.

Quality

Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected IL-23R on 10 µg of Jurkat cell lysate.

Target description

~42 kDa Observed.
May detect other isoforms of IL-23R produced by alternative splicing: isoform 1 (~72 kDa), isoform 2 (~43 kDa), isoform 3 (~41 kDa), isoform 4 (~20 kDa), isoform 5 (~30 kDa), isoform 6 (~43 kDa) and isoform 7 (~26 kDa).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moyu Ming et al.
Molecular medicine reports, 15(2), 777-783 (2016-12-31)
Allergic asthma is a chronic airway disorder characterized by airway inflammation, mucus hypersecretion, and airway hyperresponsiveness (AHR). A murine model of asthma was used to examine the antiasthmatic effect of inhaled inactived Mycobacterium phlei (M. phlei). AHR, neutrophil levels, eosinophil levels and
Yeast ubiquitin-like genes are involved in duplication of the microtubule organizing center.
S Biggins,I Ivanovska,M D Rose
The Journal of cell biology null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service