Skip to Content
Merck
All Photos(2)

Key Documents

06-1289

Sigma-Aldrich

Anti-Cyclin T1 Antibody

serum, from sheep

Synonym(s):

CDK9-associated C-type protein, Human immunodeficiency virus-1 expression, cyclin C-related protein, cyclin T1, cyclin T1b, human immunodeficiency virus type 1 (HIV-1) expression (elevated) 1, subunit of positive elongation transcription factor b

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€316.00

€316.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€316.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€316.00


Estimated to ship on16 May 2025


Request a Bulk Order

biological source

sheep

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CCNT1(904)
mouse ... Ccnt1(12455)

General description

Cyclin T1, which is also known as CCNT, is a member of the cyclin family and functions as a CDK kinase regulator. Cyclin T1 is one of regulatory cyclins (T1, T2a, T2b, and K) that is closely associated with CDK9 kinase forming a heterodimer called P-TEFb. This kinase complex is involved in phosporylating the carboxyl-terminal domain (CTD) of the large RNA polymerase II subunit which stimulate the elongation upon initiation. The cyclin T1/CDK9 complex has also been referred to as tat-associated kinase. Tat is an HIV protein that targets the cyclin T1/CDK9 complex to activate the transcription of the virus.

Specificity

Rat (85% sequence homology).
This antibody recognizes Cyclin T1.

Immunogen

Epitope: Unknown
His-tagged recombinant protein corresponding to human Cyclin T1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use Anti-Cyclin T1 Antibody (Sheep Polyclonal Antibody) validated in WB to detect Cyclin T1 also known as CDK9-associated C-type protein, cyclin C-related protein.
Western Blot Analysis: A 1:1000 dilution from a previous lot detected Cyclin T1 in 10 µg of HEK293, HeLa, HepG2, Jurkat, and PC3 cell lysates.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: A 1:1000 dilution of this antibody detected Cyclin T1 in 10 µg of HeLa cell lysate.

Target description

Cyclin T1 is expected to be ~85 kDa This antibody detects a ~85 kDa band in most cell lines. It also detects the degraded forms of Cyclin T1 (~70 kDa, ~60 kDa, et al), which is beneficial for IP applications. According to independent laboratory, this antibody can immunoprecipitate multiple forms of Cyclin T1.

Physical form

Sheep polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mechanism of release of P-TEFb and HEXIM1 from the 7SK snRNP by viral and cellular activators includes a conformational change in 7SK.
Krueger, BJ; Varzavand, K; Cooper, JJ; Price, DH
Testing null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service