Skip to Content
Merck
All Photos(1)

Key Documents

05-881

Sigma-Aldrich

Anti-Tissue Factor Antibody, clone GMA-320

clone GMA-320, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€306.00

€306.00


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€306.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€306.00


Estimated to ship on15 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

GMA-320, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... F3(2152)

Specificity

tissue factor

Immunogen

Purified human tissue factor

Application

Anti-Tissue Factor Antibody, clone GMA-320 detects level of Tissue Factor & has been published & validated for use in ELISA & WB.

Quality

routinely evaluated by immunoblot on human tissue factor

Target description

Mr 33kDa

Physical form

Format: Purified

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Cugno et al.
PloS one, 9(11), e111862-e111862 (2014-11-07)
Thrombotic risk is increased in eosinophil-mediated disorders, and several hypotheses have been proposed to link eosinophilia and thrombosis. In particular, eosinophils have been described as source of tissue factor (TF), the main initiator of blood coagulation; however, this aspect is
Effect of heparanase inhibitor on tissue factor overexpression in platelets and endothelial cells induced by anti-β2-GPI antibodies: Reply to comment from Mackman et al.
Antonella Capozzi et al.
Journal of thrombosis and haemostasis : JTH, 20(1), 261-262 (2021-12-27)
Bacterial lipoprotein TLR2 agonists broadly modulate endothelial function and coagulation pathways in vitro and in vivo.
Shin, HS; Xu, F; Bagchi, A; Herrup, E; Prakash, A; Valentine, C; Kulkarni, H; Wilhelmsen et al.
Journal of immunology (Baltimore, Md. : 1950) (1950)
Vascular biology-the role of tissue factor.
Hathcock, James
Seminars in Hematology, 41, 30-34 (2004)
Targeting tissue factor as an antithrombotic strategy.
Golino, Paolo and Cimmino, Giovanni
Seminars in Vascular Medicine, 3, 205-214 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service