Skip to Content
Merck
All Photos(2)

Documents

05-804

Sigma-Aldrich

Anti-Tau (4-repeat isoform RD4) Antibody, clone 1E1/A6

culture supernatant, clone 1E1/A6, Upstate®

Synonym(s):

Anti-Tau antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

1E1/A6, monoclonal

species reactivity

mouse, bovine, human, rat

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Microtubule Associated Proteins, or MAPS, bind to the tubulin subunits of microtubule structures and regulate their functional stability. In the cell MAPs bind to monomer and multimerized tubulin. MAP binding to multimerized tubulin further stabilizes the formation of higher order microtubulin structures. MAP binding to microtubule structures is mediated through phosporylation through Microtubule Affinity Regulated Kinase (MARK). Phosphorylation releases MAPs bound to microtubules, destabilizing the structure, driving it toward disassembly. There are predominately two MAP types, I, II. Type II MAP includes MAP2, MAP4, and tau and are found in nervous tissue. Six tau isoforms exist in brain tissue, and they are distinguished by their number of binding domains. Three isoforms have three binding domains and the other three have four binding domains. The binding domains are located in the carboxy-terminus of the protein and are positively-charged (allowing it to bind to the negatively-charged microtubule). The isoforms with four binding domains are better at stabilizing microtubules than those with three binding domains.

Specificity

Cross-reactivity with mouse and bovine expected due to sequence homology.
Recognizes Tau (4-repeat isoform RD4), Mr 45-65 kDa. Higher MW band (68-72 kDa) represents phosphorylated Tau.

Immunogen

Bovine thyroglobulin-conjugated synthetic peptide corresponding to amino acids 275-291 (VQIINKKLDLSNVQSKC) of human Tau (4-repeat isoform RD4). Region is flanking junction coded by adjacent exons 9 & 11 with the inclusion of exon 10. The immunizing sequence is identical in human, rat, mouse and bovine.

Application

Immunohistochemistry:
This antibody has been reported by an independent laboratory to detect Tau (4-repeat isoform RD4) in autoclaved paraffin brain sections. (De Silva, R., 2003;Togo, T., 2002)

Differential Detection of Tauopathies: (Togo, T., 2002)
Research Category
Neuroscience
Research Sub Category
Kinases & Phosphatases

Neurodegenerative Diseases
This Anti-Tau (4-repeat isoform RD4) Antibody, clone 1E1/A6 is validated for use in IH, WB for the detection of Tau (4-repeat isoform RD4).

Quality

Routinely evaluated by western blot on rat brain cytosol fraction.

Western Blot Analysis:
A 1:1,000-1:10,000 dilution of this lot detected Tau (4-repeat isoform RD4) in a rat brain cytosolic fraction.

Target description

~68-72 kDa

Physical form

Mouse monoclonal cultured supernatant containing 0.05% sodium azide.
Frozen at -20°C.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Rat brain cytosolic fraction.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pathological inclusion bodies in tauopathies contain distinct complements of tau with three or four microtubule-binding repeat domains as demonstrated by new specific monoclonal antibodies.
de Silva, R, et al.
Neuropathology and Applied Neurobiology, 29, 288-302 (2003)
The complex relationship between soluble and insoluble tau in tauopathies revealed by efficient dephosphorylation and specific antibodies
Hanger, D P, et al
Febs Letters, 531, 538-542 (2002)
Parenchymal and vascular lesions in ageing equine brains: histological and immunohistochemical studies.
M T Capucchio,M Marquez,P Pregel,L Foradada,M Bravo,G Mattutino,C Torre,D Schiffer et al.
Journal of Comparative Pathology null
Pamela McMillan et al.
The Journal of comparative neurology, 511(6), 788-803 (2008-10-18)
Tau is a microtubule-associated protein implicated in neurodegenerative tauopathies. Alternative splicing of the tau gene (MAPT) generates six tau isoforms, distinguishable by the exclusion or inclusion of a repeat region of exon 10, which are referred to as 3-repeat (3R)
Marion Hogg et al.
Acta neuropathologica, 106(4), 323-336 (2003-07-29)
We report a case of rapidly progressive frontotemporal dementia presenting at age 33 years. At autopsy there was severe atrophy of the frontal and temporal lobes. Tau-positive Pick bodies, which ultrastructurally were composed of straight filaments, were present, accompanied by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service