Skip to Content
Merck
All Photos(1)

Key Documents

05-682

Sigma-Aldrich

Anti-Mps1 Antibody, NT, clone 3-472-1

clone 3-472-1, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€515.00

€515.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
€515.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€515.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3-472-1, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... C5AR2(27202)

General description

Mps1 was previously known as TTK/PYK

Application

Detect Mps1 with Anti-Mps1 Antibody, NT, clone 3-472-1 (Mouse Monoclonal Antibody), that has been shown to work in WB.

Quality

routinely evaluated by immunoblot on HeLa total cell extract

Target description

97kDa

Physical form

Format: Purified

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mandar R Bhonde et al.
The Journal of biological chemistry, 281(13), 8675-8685 (2006-02-01)
DNA damage induced by the topoisomerase I inhibitor irinotecan (CPT-11) triggers in p53(WT) colorectal carcinoma cells a long term cell cycle arrest and in p53MUT cells a transient arrest followed by apoptosis (Magrini, R., Bhonde, M. R., Hanski, M. L.
Christopher P Mattison et al.
Cell cycle (Georgetown, Tex.), 10(5), 783-793 (2011-02-18)
The Mps1 family of protein kinases contributes to cell cycle control by regulating multiple microtubule cytoskeleton activities. We have uncovered a new Mps1 substrate that provides a novel link between Mps1 and the actin cytoskeleton. We have identified a conserved
Elodie Montaudon et al.
Nature communications, 11(1), 4053-4053 (2020-08-15)
A significant proportion of patients with oestrogen receptor (ER) positive breast cancers (BC) develop resistance to endocrine treatments (ET) and relapse with metastatic disease. Here we perform whole exome sequencing and gene expression analysis of matched primary breast tumours and
Recruitment of Mad1 to metaphase kinetochores is sufficient to reactivate the mitotic checkpoint.
Ballister, ER; Riegman, M; Lampson, MA
The Journal of cell biology null
Pingzhao Zhang et al.
Oncotarget, 6(39), 41550-41565 (2015-11-26)
Regulated interactions between kinetochores and spindle microtubules are critical for maintaining genomic stability during chromosome segregation. Defects in chromosome segregation are widespread phenomenon in human cancers that are thought to serve as the fuel for tumorigenic progression. Tumor suppressor proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service