Skip to Content
Merck
All Photos(1)

Key Documents

05-579

Sigma-Aldrich

Anti-Endostatin Antibody, clone 1837.46

clone 1837.46, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€484.00

€484.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
€484.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€484.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1837.46, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Col18A1(12822)

Specificity

endostatin

Immunogen

Full-length recombinant murine endostatin

Application

Anti-Endostatin Antibody, clone 1837.46 is an antibody against Endostatin for use in WB, IH.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

routinely evaluated by immunoblot on 3T3 cell lysates

Target description

Recognizes endostatin, Mr 22kDa An unidentified protein is also weakly detected at Mr 54kDa

Physical form

50mM sodium phosphate, pH 8.0, 0.05% sodium azide, before the addition of glycerol to 30%
Format: Purified
Thiophilic chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B V Sauter et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(9), 4802-4807 (2000-04-12)
Inhibition of angiogenesis has been shown to be an effective strategy in cancer therapy in mice. However, its widespread application has been hampered by difficulties in the large-scale production of the antiangiogenic proteins. This limitation may be resolved by in
M C Cichy et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 42(12), 1150-1155 (2009-11-07)
Acute renal failure (ARF) is a frequent complication of Gram-negative sepsis, with a high risk of mortality. Lipopolysaccharide (LPS)-induced ARF is associated with hemodynamic changes that are strongly influenced by the overproduction of nitric oxide (NO) through the cytokine-mediated up-regulation
H M Strik et al.
Cancer, 91(5), 1013-1019 (2001-03-17)
Endostatin is a potent inhibitor of endothelial cell proliferation, angiogenesis, and tumor growth. Its occurrence and localization has not yet been examined in human brain tumors. The authors report the production of a monoclonal antibody and detection of endostatin in
M Dhanabal et al.
The Journal of biological chemistry, 274(17), 11721-11726 (1999-04-17)
Endostatin, a carboxyl-terminal fragment of collagen XVIII, has been shown to regress tumors in mice. In this study, we have analyzed the mechanism of endostatin action on endothelial cells and nonendothelial cells. Endostatin treatment of cow pulmonary artery endothelial cells
Induction of the RNA regulator LIN28A is required for the growth and pathogenesis of RESTless breast tumors.
Gunsalus, KT; Wagoner, MP; Meyer, K; Potter, WB; Schoenike, B; Kim, S; Alexander et al.
Cancer Research null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service