Skip to Content
Merck
All Photos(1)

Key Documents

05-545

Sigma-Aldrich

Anti-PP2A Antibody, C subunit, clone 7A6

culture supernatant, clone 7A6, Upstate®

Synonym(s):

Anti-NEDLBA, Anti-PP2CA, Anti-PP2Calpha, Anti-RP-C

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€475.00

€475.00


Estimated to ship on21 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1154

Request a Bulk Order

Select a Size

Change View
200 μL
€475.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€475.00


Estimated to ship on21 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1154

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

7A6, monoclonal

species reactivity

mouse, Saccharomyces cerevisiae, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PPP2CA(5515)

Specificity

Recognizes the catalytic subunit of PP2A.

Immunogen

peptide (C-bARRTPDYFL) corresponding to amino acids 302-309 of human PP2A catalytic subunit

Application

Anti-PP2A Antibody, C subunit, clone 7A6 detects level of PP2A & has been published & validated for use in IP & WB.
Research Category
Signaling

Neuroscience
Research Sub Category
Kinases & Phosphatases

Neurodegenerative Diseases

Quality

routinely evaluated by immunoblot on RIPA lysates from A431 cells

Target description

36 kDa

Physical form

Cell culture supernatant containing 30% glycerol and 0.05% sodium azide as a preservative.
Unpurified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of L-type calcium channel and delayed rectifier potassium channel activity by p21-activated kinase-1 in guinea pig sinoatrial node pacemaker cells.
Ke, Y; Lei, M; Collins, TP; Rakovic, S; Mattick, PA; Yamasaki, M; Brodie, MS; Terrar, DA; Solaro, RJ
Circulation Research null
Loss of protein phosphatase 2A expression correlates with phosphorylation of DP-1 and reversal of dysplasia through differentiation in a conditional mouse model of cancer progression.
Maddalena T Tilli, Shawnte L Hudgins, M Silvina Frech, Ewa D Halama, Jean-Pierre Renou et al.
Cancer Research null
Chang Y Guo et al.
The Journal of biological chemistry, 277(7), 4839-4844 (2001-11-28)
Ionizing radiation (IR) is known to activate multiple cell cycle checkpoints that are thought to enhance the ability of cells to respond to DNA damage. Protein phosphatase 2A (PP2A) has been implicated in IR-induced activation of checkpoints; therefore, Jurkat cells
Modeling initiation of Ewing sarcoma in human neural crest cells.
von Levetzow, C; Jiang, X; Gwye, Y; von Levetzow, G; Hung, L; Cooper, A; Hsu, JH; Lawlor, ER
Testing null
A Fischer et al.
Toxicology and applied pharmacology, 245(1), 9-20 (2010-02-23)
Cellular uptake of microcystins (MCs), a family of cyclic cyanobacterial heptapeptide toxins, occurs via specific organic anion transporting polypeptides (OATPs), where MCs inhibit serine/threonine-specific protein phosphatase (PP). Despite comparable PP-inhibitory capacity, MCs differ greatly in their acute toxicity, thus raising

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service