Skip to Content
Merck

Skip To

05-366

Anti-PLCγ-1 Antibody, clone B-6-4

clone B-6-4, 1 mg/mL, Upstate®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 438,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
B-6-4, monoclonal
Application:
IP, WB
Citations:
28

€ 438,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1774

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

B-6-4, monoclonal

species reactivity

human, rabbit, rat, bovine, mouse

manufacturer/tradename

Upstate®

concentration

1 mg/mL

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PLCG1(5335)

General description

~135 kDa

Immunogen

Purified soluble phospholipase C (PLC) g-1 from bovine brain

Application

Anti-PLCγ-1 Antibody, clone B-6-4 is an antibody against PLCγ-1 for use in IP & WB.
Research Category
Signaling
Research Sub Category
Lipid Signaling

Biochem/physiol Actions

PLCγ-1; may also react with some non-PLC γ-1 SH-3 containing proteins

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1 antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.
Routinely evaluated in immunoblot using acid precipitated protein from bovine brain, RIPA lysates from human A431 cells, and previously using mouse 3T3/A31 fibroblasts and rat L6 myoblasts lysates.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
05-78007-383MAB012
clone

B-6-4, monoclonal

clone

GMA104, monoclonal

clone

polyclonal

clone

B1, monoclonal

species reactivity

human, rabbit, rat, bovine, mouse

species reactivity

human

species reactivity

human

species reactivity

human

antibody form

purified immunoglobulin

antibody form

purified antibody

antibody form

affinity purified immunoglobulin

antibody form

purified antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

mouse

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

ELISA: suitable, radioimmunoassay: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters