Skip to Content
Merck
All Photos(4)

Key Documents

05-235

Sigma-Aldrich

Anti-EF1α Antibody, clone CBP-KK1

clone CBP-KK1, Upstate®, from mouse

Synonym(s):

CTCL tumor antigen, EF1a-like protein, Elongation factor 1 A-1, Elongation factor Tu, Leukocyte receptor cluster member 7, cervical cancer suppressor 3, elongation factor 1 alpha subunit, elongation factor 1-alpha, eukaryotic translation elongation facto

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€528.00

€528.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
€528.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€528.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CBP-KK1, monoclonal

species reactivity

bacteria, Saccharomyces cerevisiae

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... EEF1A1(1915)

General description

Human eukaryotic translation elongation factor 1 alpha has two isoforms, alpha 1 and alpha 2. The alpha 1 isoform gene encodes a subunit of the elongation factor-1 complex which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. The alpha 1 isoform is expressed in brain, placenta, lung, liver, kidney, and pancreas. The other isoform (alpha 2) is expressed in brain, heart and skeletal muscle.
The alpha 1 isoform is identified as an autoantigen in 66% of patients with Felty syndrome. The gene for the alpha 1 isoform has been found to have multiple copies on many chromosomes, some of which, if not all, represent different pseudogenes. The gene for the alpha 2 may be critical in the development of ovarian cancer.

Specificity

This antibody recognizes EF-1α (Elongation factor-1 alpha), Mr 53 kDa. Occasionally, a 38 kDa protein is also detected in A431 cell lysate.

Immunogen

Crude calmodulin-binding proteins from Trypanosoma brucei isolated using calmodulin-affinity chromatography. Clone CBP-KK1.

Application

Detect EF1α using this Anti-EF1α Antibody, clone CBP-KK1 validated for use in IP & WB.
Immunoprecipitation:
4 µg of a previous lot immunoprecipitated EF1α from an A431 RIPA cell lysate.
Research Category
Protein Trafficking
Research Sub Category
RNA Metabolism & Binding Proteins

RNA Binding Protein (RBP)

Quality

Routinely evaluated on RIPA lysates from non-stimulated A431 cells, human Jurkat or mouse 3T3/A31.

Western Blot Analysis:
0.5-2 µg/mL of this lot detected EF1α in RIPA lysates from non-stimulated A431 cells. Previous lots detected EF1α in human Jurkat and mouse 3T3/A31 RIPA cell lysates.

Target description

53 kDa

Physical form

Format: Purified
Protein G Chromatography
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20ºC. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CDK2 catalytic activity and loss of nuclear tethering of retinoblastoma protein in childhood acute lymphoblastic leukemia.
Schmitz, NM; Leibundgut, K; Hirt, A
Leukemia null
Molecular determinants for the tissue specificity of SERMs
Shang, Y. and Brown, M.
Science (New York, N.Y.), 295, 2465-2468 (2002)
Co-infection with the friend retrovirus and mouse scrapie does not alter prion disease pathogenesis in susceptible mice.
Leblanc, P; Hasenkrug, K; Ward, A; Myers, L; Messer, RJ; Alais, S; Timmes, A; Priola, SA; Priola, S
Testing null
S R Kimball et al.
The Journal of biological chemistry, 274(17), 11647-11652 (1999-04-17)
Regulation of translation of mRNAs coding for specific proteins plays an important role in controlling cell growth, differentiation, and transformation. Two proteins have been implicated in the regulation of specific mRNA translation: eukaryotic initiation factor eIF4E and ribosomal protein S6.
ADAM9 up-regulates N-cadherin via miR-218 suppression in lung adenocarcinoma cells.
Sher, YP; Wang, LJ; Chuang, LL; Tsai, MH; Kuo, TT; Huang, CC; Chuang, EY; Lai, LC
Testing null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service