Skip to Content
Merck
All Photos(1)

Key Documents

05-23-2404

Sigma-Aldrich

Calcitonin Gene-Related Peptide-I, Human

Calcitonin Gene-Related Peptide-I, Human, CAS 90954-53-3, is a potent and long-acting vasodilator. Increases plasma levels of pancreatic polypeptide. Has chemotactic activity towards eosinophils.

Synonym(s):

Calcitonin Gene-Related Peptide-I, Human, CGRP-I, αCGRP, ACDTATCVTHRLAGLLSRSGGVVKNNFVPTNVGSKAF-NH₂

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€553.00

€553.00


Check Cart for Availability


Select a Size

Change View
1 MG
€553.00

About This Item

Empirical Formula (Hill Notation):
C163H267N51O49S2
CAS Number:
Molecular Weight:
3789.31
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

€553.00


Check Cart for Availability

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

5% acetic acid: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C163H269N51O49S2/c1-73(2)52-97(186-116(226)65-179-131(233)82(18)183-139(241)98(53-74(3)4)193-137(239)94(44-35-49-176-162(171)172)188-142(244)101(57-91-62-175-72-182-91)199-159(261)128(88(24)221)213-156(258)123(79(13)14)207-151(253)110(71-265)204-160(262)126(86(22)219)210-133(235)84(20)185-157(259)125(85(21)218)211-147(249)105(61-119(229)230)198-150(252)109(70-264)203-130(232)81(17)166)140(242)194-99(54-75(5)6)141(243)202-108(69-217)149(251)190-95(45-36-50-177-163(173)174)138(240)201-106(67-215)134(236)180-63-115(225)178-64-118(228)205-121(77(9)10)155(257)208-122(78(11)12)154(256)191-93(43-32-34-48-165)136(238)196-102(58-112(167)222)144(246)197-103(59-113(168)223)143(245)195-100(56-90-40-29-26-30-41-90)145(247)209-124(80(15)16)161(263)214-51-37-46-111(214)152(254)212-127(87(23)220)158(260)200-104(60-114(169)224)146(248)206-120(76(7)8)153(255)181-66-117(227)187-107(68-216)148(250)189-92(42-31-33-47-164)135(237)184-83(19)132(234)192-96(129(170)231)55-89-38-27-25-28-39-89/h25-30,38-41,62,72-88,92-111,120-128,215-221,264-265H,31-37,42-61,63-71,164-166H2,1-24H3,(H2,167,222)(H2,168,223)(H2,169,224)(H2,170,231)(H,175,182)(H,178,225)(H,179,233)(H,180,236)(H,181,255)(H,183,241)(H,184,237)(H,185,259)(H,186,226)(H,187,227)(H,188,244)(H,189,250)(H,190,251)(H,191,256)(H,192,234)(H,193,239)(H,194,242)(H,195,245)(H,196,238)(H,197,246)(H,198,252)(H,199,261)(H,200,260)(H,201,240)(H,202,243)(H,203,232)(H,204,262)(H,205,228)(H,206,248)(H,207,253)(H,208,257)(H,209,247)(H,210,235)(H,211,249)(H,212,254)(H,213,258)(H,229,230)(H4,171,172,176)(H4,173,174,177)/t81-,82-,83-,84-,85+,86+,87+,88+,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,120-,121-,122-,123-,124-,125-,126-,127-,128-/m0/s1

InChI key

DNKYDHSONDSTNJ-XJVRLEFXSA-N

General description

Potent and long-acting vasodilator. CGRP receptors are present in pancreatic β-cells. Increases plasma levels of pancreatic polypeptide. Has chemotactic activity towards eosinophils.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Potent and long-acting vasodilator
Product does not compete with ATP.
Reversible: no

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Ala-Cys²-Asp-Thr-Ala-Thr-Cys⁷-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asn-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Lys-Ala-Phe-NH₂ (disulfide bond: 2 → 7)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month -20°C.

Other Notes

Edwards, A.V., and Bloom, S.R. 1994. Am. J. Physiol. 267, E847.
Henke, H., et al. 1987. Brain Res.410, 404.
Chiba, T., et al. 1984. Nature 308, 746.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service