Skip to Content
Merck
All Photos(1)

Key Documents

05-170

Sigma-Aldrich

Anti-IL-2 Rα Antibody, clone 7G7/B6

clone 7G7/B6, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€461.00

€461.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
€461.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€461.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7G7/B6, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

dot blot: suitable
immunocytochemistry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... IL2RA(3559)

Specificity

Recognizes interleukin-2 receptor.

Immunogen

Whole cells expressing human IL-2 Receptor α

Application

Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
This Anti-IL-2 Rα Antibody, clone 7G7/B6 is validated for use in DB, IC for the detection of IL-2 Rα.

Quality

routinely evaluated on ice cold ethanol/acetic acid [95:5] fixed phytohemagglutinin (PHA)-treated Jurkat T lymphocytes

Target description

55 kDa

Physical form

Format: Purified
Protein G Purified
Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide as a preservative.

Storage and Stability

Lyophilized: 2 years at -20°C; Rehydrated: 6 months at -20°C

Analysis Note

Control
Lung tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A monoclonal antibody 7G7/B6, binds to an epitope on the human interleukin-2 (IL-2) receptor that is distinct from that recognized by IL-2 or anti-Tac.
Rubin, L A, et al.
Hybridoma, 4, 91-102 (1985)
Benjamin C Jennings et al.
FEBS letters, 595(13), 1758-1767 (2021-05-16)
The SARS-CoV-2 spike glycoprotein (spike) mediates viral entry by binding ACE2 receptors on host cell surfaces. Spike glycan processing and cleavage, which occur in the Golgi network, are important for fusion at the plasma membrane, promoting both virion infectivity and
P D Tahiliani et al.
The Journal of biological chemistry, 272(12), 7892-7898 (1997-03-21)
Integrin-mediated adhesion of cells to extracellular matrix proteins triggers a variety of intracellular signaling pathways including a cascade of tyrosine phosphorylations. In many cell types, the cytoplasmic focal adhesion tyrosine kinase, FAK, appears to be the initial protein that becomes
Sabine S Lange et al.
PLoS genetics, 12(1), e1005759-e1005759 (2016-01-05)
DNA polymerase ζ (pol ζ) is exceptionally important for maintaining genome stability. Inactivation of the Rev3l gene encoding the polymerase catalytic subunit causes a high frequency of chromosomal breaks, followed by lethality in mouse embryos and in primary cells. Yet
K S Lai et al.
The Journal of biological chemistry, 270(42), 25028-25036 (1995-10-20)
Signal transduction of cytokine receptors is mediated by the JAK family of tyrosine kinases. Recently, the kinase partners for the interleukin (IL)-2 receptor have been identified as JAK1 and JAK3. In this study, we report the identification of splice variants

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service