Skip to Content
Merck
All Photos(1)

Documents

05-1355

Sigma-Aldrich

Anti-acetyl Histone H4 (Lys5/8/12/16) Antibody, clone 3HH4-4C10

ascites fluid, clone 3HH4-4C10, from mouse

Synonym(s):

H4KAc, Histone H4 (acetyl), H4 histone family, member N, H4 histone, family 2, histone 2, H4, histone 2, H4a, histone IV, family 2, histone cluster 2, H4a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3HH4-4C10, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

acetylation (Lys5/Lys8/Lys12/Lys16)

Gene Information

human ... H4C1(8359)
mouse ... H4C1(326619)

General description

Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs). Posttranslational acetylation of the conserved core histone N-terminal tail domains is typically associated with gene activation.

Specificity

This antibody recognizes Histone H4.

Immunogen

Epitope: Lys5/8/12/16
Linear peptide corresponding to human Histone H4 acetylated at Lys5/8/12/16.

Application

Immunocytochemistry Analysis:
1:500 dilution from a representative lot detected Histone H4 in A431 and HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-acetyl Histone H4 (Lys5/8/12/16) Antibody, clone 3HH4-4C10 (mouse monoclonal antibody) validated in WB, ICC to detect acetyl Histone H4 (Lys5/8/12/16) also known as H4KAc, Histone H4 (acetyl).

Quality

Evaluated by Western Blot in HeLa acid extract lysate.

Western Blot Analysis: 1:2,000 dilution of this antibody detected Histone H4 on 10 µg of HeLa acid extract lysate.

Target description

~ 13 kDa

Physical form

Unpurified
Unpurified mouse monoclonal IgG1κ preservative-free ascites.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa acid extract lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingwen Feng et al.
Scientific reports, 7(1), 9581-9581 (2017-08-31)
High-content screening is commonly used in studies of the DNA damage response. The double-strand break (DSB) is one of the most harmful types of DNA damage lesions. The conventional method used to quantify DSBs is γH2AX foci counting, which requires
Madhumanti Barman et al.
Frontiers in microbiology, 12, 759817-759817 (2021-12-07)
The apicomplexan parasite, Theileria annulata, is the most prevalent hemoprotozoan in livestock, causing significant economic losses worldwide. It is essential to develop new and improved therapeutics, as current control measures are compromised by the development of resistance against the only
Yolanda Colino-Sanguino et al.
iScience, 21, 773-788 (2019-11-16)
Acetylation of the histone variant H2A.Z (H2A.Zac) occurs at active regulatory regions associated with gene expression. Although the Tip60 complex is proposed to acetylate H2A.Z, functional studies suggest additional enzymes are involved. Here, we show that p300 acetylates H2A.Z at
William Newhard et al.
SLAS discovery : advancing life sciences R & D, 26(5), 642-654 (2021-01-12)
Antiretroviral therapy is able to effectively control but not eradicate HIV infection, which can persist, leading to the need for lifelong therapy. The existence of latently HIV-infected cells is a major barrier to the eradication of chronic HIV infection. Histone
Wei Wei et al.
Cell research, 27(7), 898-915 (2017-05-13)
Recent studies on enzymes and reader proteins for histone crotonylation support a function of histone crotonylation in transcription. However, the enzyme(s) responsible for histone decrotonylation (HDCR) remains poorly defined. Moreover, it remains to be determined if histone crotonylation is physiologically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service