Skip to Content
Merck
All Photos(2)

Key Documents

04-968

Sigma-Aldrich

Anti-Folr4 (Folate receptor 4) Antibody, clone 5E12.2

clone 5E12.2, from mouse

Synonym(s):

folate receptor 4 (delta) homolog (mouse), folate receptor 4 (delta) homolog, Folate receptor 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€395.00

€395.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€395.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€395.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5E12.2, monoclonal

species reactivity

human, mouse

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Folates are necessary and critical components in the synthesis of proteins and nucleic acids, and have putative roles in the development of both the spinal canal and brain in the early stages of pregnancy (Yamaguchi, 2007). Folate receptor 4 (Folr4) is a recently discovered cell surface receptor for folic acid on T regulatory cells (Tregs). Studies have shown that Folr4 has an integral role in the maintenance of Treg cell function (Arase, 2009). Expression of Folr4 in Tregs is notably high, making it an opportune marker for the study of Treg cells. When studied in conjunction with CD4 and CD25, Folr4 also provides researchers with a marker to distinguish between Tregs and other T cells (Chai, 2008). Tregs are of interest in therapeutic, drug research due to their ability to actively suppress immune responses.

Specificity

This antibody recognizes Folate receptor 4 (Folr4).

Immunogen

Epitope: Unknown
KLH-conjugated linear peptide corresponding to human Folate receptor 4 (Folr4).

Application

Anti-Folr4 (Folate receptor 4) Antibody, clone 5E12.2 detects level of Folr4 (Folate receptor 4) & has been published & validated for use in WB.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
Western Blot (SNAP ID) Analysis: 1 µg/mL of this antibody detected Folate receptor 4 (Folr4) in 10 µg of human thymus tissue lysate.

Quality

Evaluated by Western Blot in human thymus tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Folate receptor 4 (Folr4) in 10 µg of human thymus tissue lysate.
A previous lot was shown to detect Folate Receptor 4 in mouse thymus tissue lysate.

Target description

~26 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human thymus tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service