Skip to Content
Merck
All Photos(1)

Key Documents

04-903

Sigma-Aldrich

Anti-GSK3α/β Antibody, rabbit monoclonal

culture supernatant, from rabbit

Synonym(s):

GSK-3 alpha, glycogen synthase kinase 3 alpha

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€408.00

€408.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

monoclonal

species reactivity

mouse, human, rat

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GSK3B(2932)

General description

Glycogen Synthase Kinase 3 differs from most serine/threonine kinases in that it is active in the absence of the action of signaling pathways. Two isoforms exist, GSK3α and GSK3β. The function of GSK3 is to phosphorylate Glycogen Synthase and thereby inactivate it. Insulin action stimulates the PI3 Kinase pathway, resulting in Akt activation, which phosphorylates and inactivates GSK3. Glycogen Synthase is then rapidly dephosphorylated and activated. Other GSK3 substrates include Jun (on inhibitory sites), and eIF2B. Phosphorylation of Tau by GSK3 may relate to development of Alzheimer′s disease. Phosphorylation state-specific antibodies directed against the Akt site (Ser21) on GSK3 are suitable for surrogate assays of the activation state of the pathway.

Specificity

GSK3 alpha and beta.
Predicted to cross-react with rat based on sequence homology.

Immunogen

N-terminal 6His-tagged recombinant full length human GSK3β.

Application

Immunoprecipitation: 2-8 μL of a previous lot of antibody immunoprecipitated GSK3 alpha and beta from 100 μg of HEK293 RIPA lysate.
NOTE: This antibody also successfully immunoprecipitated GSK3 alpha and beta using the Catch and Release v2.0 Reversible Immunoprecipitation System (Catalog # 17-500).
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling
This Anti-GSK3α/β Antibody is validated for use in WB, IP for the detection of GSK3α/β.

Quality

Evaluated by western blot on lysates from HEK293 cells.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected GSK3 alpha and GSK3 beta in RIPA lysates from A431, HEK293, Jurkat, HeLa NE or 3T3/A31 cells.

Target description

~46 kDa (beta) and 51 kDa (alpha)

Linkage

Replaces: 05-903

Physical form

Cultured supernantant containing 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes Menzel-Severing et al.
Scientific reports, 8(1), 10268-10268 (2018-07-08)
Understanding transcription factor (TF) regulation of limbal epithelial stem/progenitor cells (LEPCs) may aid in using non-ocular cells to regenerate the corneal surface. This study aimed to identify and characterize TF genes expressed specifically in LEPCs isolated from human donor eyes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service