Skip to Content
Merck
All Photos(2)

Key Documents

04-804

Sigma-Aldrich

Anti-NFAT1 Antibody, clone 8G4.2

clone 8G4.2, Upstate®, from mouse

Synonym(s):

Nuclear factor of activated T-cells, cytoplasmic 2, NF-ATc2, NFATc2, NFAT pre-existing subunit, NF-Atp, T-cell transcription factor NFAT1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€358.00

€358.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€358.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€358.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8G4.2, monoclonal

species reactivity

rat, mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NFATC2(4773)

General description

NFAT (Nuclear Factor of Activated T-cells) is a family of transcription factors that are located in the cytoplasm in their inactive state. NFAT translocates to the nucleus upon stimulation of the T-cell receptor where it plays a key role in gene expression in immune response. NFAT is dephosphorylated and activated by the phosphatase PP2B/Calcineurin. This results in its nuclear localization and transcriptional activity. For NFAT1 (NFATC2, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2), calcineurin is known to target the region between residues 114-126. Mutations in this region of NFAT1 result in its inability to de-phosphorylate and subsequent nuclear translocation. Upon activation, NFAT mediates the transcriptional induction of cytokines and other immuno-regulatory genes during immune responses, and is expressed in T-cells, B-cells, mast cells and NK cells.

Specificity

Detects NFAT1.
Other species have not been tested.

Immunogen

Epitope: N-term
GST-fusion protein corresponding to amino acids 1-297 of human NFAT1.

Application

Research Category
Signaling
Research Sub Category
Transcription Factors
This Anti-NFAT1 Antibody, clone 8G4.2 is validated for use in WB, IF for the detection of NFAT1.

Quality

Routinely evaluated by Western Blot

Target description

120-140 kDa

Physical form

Format: Purified
Purified mouse monoclonal in PBS with 0.05% NaN3.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service