Skip to Content
Merck

Skip To

04-259

Anti-19S regulator non-ATPase subunit S5a/Rpn10 Antibody, clone S5a-18

clone S5a-18, Upstate®, from mouse

Synonym(s):

S5a

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
S5a-18, monoclonal
Application:
IP, WB
Citations:
34
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S5a-18, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PSMD4(5710)

General description

41 kDa
Rpn10 (S5a) is a non-ATPase subunit of the 19S regulatory complex of the 26s proteasome. Subunit Rpn10 (S5a) of the 26S proteasome was originally identified as a protein capable of binding poly-ubiquitin chains of four or more ubiquitins, in the absence of other 26S proteasome subunits.

Immunogen

Full length human recombinant Rpn10 protein.

Application

Detect 19S regulator non-ATPase subunit S5a/Rpn10 using this Anti-19S regulator non-ATPase subunit S5a/Rpn10 Antibody, clone S5a-18 validated for use in IP & WB.
Research Category
Protein Trafficking
Research Sub Category
Ubiquitin & Ubiquitin Metabolism

Biochem/physiol Actions

Recognizes 19S regulator non-ATPase subunit S5a/Rpn10.

Physical form

100 µL of purified immunoglobulin in PBS containing 0.01M sodium azide. Dilute to working strength with phosphate buffered saline pH 7.2-7.4.
Format: Purified

Preparation Note

Stable for 1 year at -20°C from date of shipment for up to 12 months.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters