Skip to Content
Merck
All Photos(1)

Key Documents

04-1467

Sigma-Aldrich

Anti-PABP 1 Antibody, clone 10E10

clone 10E10, from mouse

Synonym(s):

PABP 1, Poly(A)-binding protein 1, poly(A) binding protein, cytoplasmic 1, poly(A) binding protein, cytoplasmic 2, poly(A)-binding protein, cytoplasmic 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10E10, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PABPC1(26986)

General description

Poly(A)-binding protein (PABP 1) is an abundant protein that interacts with the 3′ poly(A) tail of most eukaryotic mRNAs. In vivo this protein is involved in many aspects of mRNA metabolism, including mRNA stability, translationally coupled mRNA turnover, protein biosynthesis, and ribosome biogenesis. PABP1 shuttles between the cytoplasm and the nucleus and may bind to cytoplasmic RNA sequences other than poly(A). PABP1 accumulates in the nucleus if transcription is inhibited, suggesting that nuclear export is coupled to active transcription.

Specificity

This antibody recognizes PABP 1.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human PABP 1.

Application

Anti-PABP 1, clone 10E10 is a Mouse Monoclonal Antibody for detection of PABP 1 also known as PABP 1, Poly(A)-binding protein 1, poly(A) binding protein cytoplasmic 1 & has been validated in WB, ICC & IP.
Immunoprecipitation:
Cited in Thoma, C., et al. (2008) RNA. 14:1579-1589, and Buchet-Poyau, K., et al.(2007) Nuc. Acids Res. 35:1289-1300 using a representative lot.

Immunocytochemistry:
Cited in Afonina, E, et al. (1998) J. Biol. Chem., 273: 13015-21 (1998) using a representative lot.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/ml of this antibody detected PABP 1 in 10 µg of HeLa cell lysate.

Target description

~ 71 kDa

Linkage

Replaces: 05-847

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgGκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideki Nakamura
Methods in molecular biology (Clifton, N.J.), 2312, 253-276 (2021-07-07)
Recent studies revealed the biological significance of dynamic multicomponent assemblies of biomolecules inside living cells. Protein and nucleic acid assemblies are biomolecular condensates or non-membrane-bound organelles that have attracted increasing attention. Synthetic tools that manipulate the dynamic assembly/disassembly process of
Rebekah Koppenol et al.
Brain : a journal of neurology, 146(6), 2346-2363 (2022-12-14)
Polyglutamine diseases are a group of neurodegenerative disorders caused by an abnormal expansion of CAG repeat tracts in the codifying regions of nine, otherwise unrelated, genes. While the protein products of these genes are suggested to play diverse cellular roles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service