Skip to Content
Merck

Skip To

04-002

Anti-SOCS1 Antibody, clone 4H1

clone 4H1, Upstate®, from mouse

Synonym(s):

suppressor of cytokine signaling 1, CIS1, CISH1, JAB, SOCS-1, SSI-1, SSI1, TIP3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 402,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
4H1, monoclonal
Application:
IP, WB
Citations:
15

€ 402,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4H1, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SOCS1(8651)

General description

37 kDa

Immunogen

mouse SOCS1 recombinant GST fusion protein encompassing amino acids 2-78.

Application

Anti-SOCS1 Antibody, clone 4H1 is an antibody against SOCS1 for use in IP & WB.
Immunoblot Analysis: 1:1,000 dilution of this antibody detected SOCS1 in KG-1 cell lysate.
Immunoprecipitation (IP): 10 μg of this antibody immunoprecipitated SOCS1 from 200 μL of cell extract in RIPA buffer.
Research Category
Signaling
Research Sub Category
Immunological Signaling

Biochem/physiol Actions

Recognizes SOCS1

Physical form

Format: Purified
Protein G Purified

Preparation Note

2 years at 2° to 8°C from date of shipment.

Analysis Note

Routinely evaluated by immunoblot.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
04-004SAB5700844PRS3767
biological source

mouse

biological source

mouse

biological source

rabbit

biological source

rabbit

species reactivity

mouse, human

species reactivity

mouse, human

species reactivity

rat, mouse, human

species reactivity

human

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

clone

4H1, monoclonal

clone

1B2, monoclonal

clone

polyclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

western blot: suitable

technique(s)

immunohistochemistry: 1:50-1:200, western blot: 1:500-1:2000

technique(s)

indirect ELISA: suitable, western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters