Skip to Content
Merck
All Photos(1)

Key Documents

03-0191-00

Millipore

SMC® Human IFN-β1 High Sensitivity Immunoassay Kit

Synonym(s):

Interferon β-1 immunoassay kit, Interferon beta-1 Single Molecule Counting Kit, Interferon beta-1 immunoassay kit, Interferon beta-1 ultra-sensitive immunoassay

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€1,000.00

€1,000.00


Estimated to ship on23 April 2025


Request more information

Select a Size

Change View
1 EACH
€1,000.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€1,000.00


Estimated to ship on23 April 2025


Request more information

manufacturer/tradename

SMC®

assay range

sensitivity: 0.15 pg/mL
(lower limit of quantification)

inter-assay cv: 13%
intra-assay cv: 6%

technique(s)

ELISA: suitable
single molecule counting: suitable

relevant disease(s)

infectious diseases; autoimmune diseases; neurodegenerative disorders

storage temp.

2-8°C

General description

A member of the type I interferon family of glycoprotein second messengers, human Interferon Beta (IFN-ß1) has been associated with antiviral activity as a secreted factor from infected cells that mediates innate immunity in neighboring cells. In addition, IFN-β1 has also been utilized as treatment for Multiple Sclerosis (MS) due to its ability to ameliorate inflammation in the central nervous system in this disease.

The SMC® Human IFN-β1 High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format.
Assay protocols are similar to existing sandwich ELISA methods with two key differences:
1) Elution buffer disrupts the sandwich, separating the labeled detection antibody for quantification.
2) The Erenna and SMCxPRO® immunoassay systems detect analytes using Single Molecule Counting (SMC®) technology.

Specificity

Cross Reactivity:
Cross-reactivity to the following analytes were tested with the following results:
  • human IFN-α2 – 0.2%
  • human IFN-γ – 0.3%
  • human IL-29 (IFN-λ1) – 0.2%
  • human IL-28A (IFN-λ2) – 0.2%
  • human IL-28B (IFN-λ3) – 0.3%
  • human IFN-ω – 0.2%

Accuracy:
  • Minimum of 5 serum samples spiked with average recovery of 88% with a range of 78-100%
  • Minimum of 5 plasma samples spiked with average recovery of 93% with a range of 75-113%

Application

Research Category:
  • Inflammation & Immunology
  • Immune Response
  • Infectious Diseases
  • Signaling
  • Neuroscience

Research Sub-Category:
  • Inflammatory Disease
  • Immune Response
  • Autoimmune disease
  • Neurodegenerative Diseases
  • Neuroregenerative Medicine
  • Virology
  • Infectious Diseases - Viral
  • Immunological Signaling

The sensitivity of the Single Molecule Counting (SMC®) technology gives the SMC® Human IFN-β1 High Sensitivity Immunoassay Kit unparalleled performance in the quantification of IFN-β1 in human serum and plasma.

Features and Benefits

This new streamline kit has the following features including:

  • Improved options for sample prep and incubation
  • Lyophilization of the standard provides the kit at one storage condition
  • Increased reagent volumes to accommodate automation
  • Validated to run on both SMCxPRO® and Erenna

Packaging

96 well plate

Components

  • Assay Buffer
  • IFN-β1 Coated Beads
  • Standard Diluent
  • IFN-β1 Detection Antibody
  • IFN-β1 Standard
  • 10X Wash Buffer
  • Buffer D
  • Elution Buffer B

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Explore ultrasensitive immunoassay technology with SMC® technology, including its biomarker detection platform, advantages of high sensitivity assays, and more.

Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service