Skip to Content
Merck
All Photos(1)

Key Documents

03-0187-00

Millipore

SMC® Human MCP-1 High Sensitivity Immunoassay Kit

Synonym(s):

MCP 1 immunoassay kit, MCP-1 Single Molecule Counting Kit, MCP-1 ultra-sensitive immunoassay, Monocyte Chemoattractant Protein-1 immunoassay kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€1,000.00

€1,000.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 KIT
€1,000.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€1,000.00


Check Cart for Availability

Request more information

manufacturer/tradename

SMC®

assay range

sensitivity: 0.15 pg/mL
(Lower Limit of Quantification)

inter-assay cv: 4%
intra-assay cv: 5%
(five normal plasma samples run in triplicate by 3 different operators on 3 different days.)

technique(s)

single molecule counting: suitable

storage temp.

2-8°C

Specificity

Species Reactivity:
Specificity to the following species analytes were tested with the following results:
Rat – Not cross-reactive
Mouse – Not cross-reactive
Canine – Not cross-reactive

Cross Reactivity:
Cross-reactivity to the following analytes were tested with the following results:
MCP-2 – Not cross-reactive
MCP-3 – Not cross-reactive

Accuracy:
  • Minimum of 5 serum samples spiked with average recovery of 104% with a range of 92-115%
  • Minimum of 5 plasma samples spiked with average recovery of 101% with a range of 80-123%

Application

The sensitivity of the Single Molecule Counting (SMC®) technology gives the SMC® Human MCP-1 High Sensitivity Immunoassay Kit unparalleled performance in the quantification of MCP-1 in human serum and plasma.

This new streamline kit has the following features including:
  • Improved options for sample prep and incubation
  • Lyophilization of the standard provides the kit at one storage condition
  • Removal of a plate transfer step
  • Increased reagent volumes to accommodate automation
  • Validated to run on both SMCxPRO® and Erenna


Research Category:

Inflammation & Immunology

Research Sub-Category:
  • Inflammatory Disease
  • Immune Response
  • Autoimmune disease

Packaging

96 well assay

Components

  • Assay Buffer
  • MCP-1 Coated Beads
  • Standard Diluent
  • MCP-1 Detection Antibody
  • MCP-1 Standard
  • 10X Wash Buffer
  • Buffer D
  • Elution Buffer B

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service