Skip to Content
Merck
All Photos(1)

Documents

03-0183-00

Millipore

SMC® Human UCHL1 High Sensitivity Immunoassay Kit

Synonym(s):

HEL-S-53, NDGOA, PARK5, PGP9.5 high sensitive immuno assay, SPG79A, Ubiquitin Carboxyl Terminal Hydrolase L1 immunoassay kit, Uch-L1 single molecule counting kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

manufacturer/tradename

SMC®

technique(s)

single molecule counting: suitable

Legal Information

SMC is a registered trademark of Singulex

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service