Skip to Content
Merck
All Photos(1)

Documents

03-0177-00

Millipore

SMC® Human IL-17A/F Heterodimer High Sensitivity Immunoassay Kit

Synonym(s):

IL-17A/F Single Molecule Counting Kit, IL-17A/F ultra-sensitive immunoassay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

species reactivity

monkey, canine, nonhuman primates

manufacturer/tradename

SMC®

assay range

sensitivity: 2.34 pg/mL
(Lower Limit of Quantification)

inter-assay cv: <20%
intra-assay cv: <15%
(Intra-assay n=20; Inter-assay n=5)

technique(s)

single molecule counting: suitable

storage temp.

2-8°C

Specificity

Species Reactivity:
Although IL-17A/F was detectable in non-human primate/monkey and canine, the assay has not been validated for these species.

Accuracy:
  • Minimum of 5 serum samples spiked with average recovery of 101% with a range of 91-114%
  • Minimum of 5 plasma samples spiked with average recovery of 88% with a range of 75-111%

Application

The sensitivity of the Single Molecule Counting (SMC®) technology gives the SMC® Human IL-17A/F High Sensitivity Immunoassay Kit unparalleled performance in the quantification of IL-17A/F in human serum and plasma.

This new streamline kit has been improved with the following features including new including new improvements over the legacy IL-17A/F kit, #03-0119-00:
  • Adjusted standard curve--changed to 0.3 – 300 pg/mL from 0.59 – 600 pg/mL
  • Improved options for sample prep and incubation
  • Lyophilization of the standard provides the kit at one storage conditi
  • Removal of a plate transfer ste• Increased reagent volumes to accommodate automation
  • Validated to run on both SMCxPRO® and Erenna


Research Category:
Inflammation & Immunology

Research Sub-Category:
  • Inflammatory Disease
  • Immune Response
  • Autoimmune disease

Packaging

96 well assay

Components

  • Assay Buffer
  • IL-17A/F Coated Beads
  • Standard Diluent
  • IL-17A/F Detection Antibody
  • IL-17A/F Standard
  • 10X Wash Buffer
  • Buffer D
  • Elution Buffer B

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Immunoassays in cosmetics research assess product safety and effects on inflammation, aging, and tissue regeneration.

Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service