Skip to Content
Merck
All Photos(1)

Documents

03-0171-00

Millipore

SMC® Human Brain-Derived Neurotrophic Factor (BDNF) High Sensitivity Immunoassay Kit

Synonym(s):

Abrineurin immunoassay kit, BDNF Single Molecule Counting Kit, BDNF ultra-sensitive immunoassay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

manufacturer/tradename

SMC®

assay range

sensitivity: 0.04 pg/mL
(lower limit of quantification)

inter-assay cv: <11%
intra-assay cv: <10%
((Intra-assay CV n=20; Inter-assay n=5))

technique(s)

ELISA: suitable
single molecule counting: suitable

relevant disease(s)

Alzheimer′s disease

storage temp.

2-8°C

General description

96 well assay
Brain-derived neurotrophic factor (BDNF) is a protein made by neuronal cells critical for the growth and functioning of neurons as well as binding at synapses to improve signal strength between neurons. While sufficient levels of BDNF promote the function and growth of brain cells and neurons, a decrease in BDNF levels can lead to impaired neurological function and is associated with diseases such as Alzheimer′s, epilepsy, schizophrenia and depression.

The SMC® Human Brain-Derived Neurotrophic Factor (BDNF) High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format.
Assay protocols are similar to existing sandwich ELISA methods with two key differences:
1) Elution buffer disrupts the sandwich, separating the labeled detection antibody for quantification.
2) The Erenna and SMCxPRO® immunoassay systems detect analytes using Single Molecule Counting (SMC®) technology.

Specificity

Cross Reactivity:
Cross reactivity and interference of ProBDNF was tested in this assay.
The assay shows 1.2% cross reactivity with ProBDNF. Serum and plasma spiked with ProBDNF show an average 4.4% interference.

Accuracy:
  • Minimum of 5 EDTA plasma samples spiked with average recovery of 96% with a range of 81-108%
  • Minimum of 5 serum samples spiked with average recovery of 90% with a range of 71-98%
  • Minimum of 5 cerebrospinal fluid (CSF) samples spiked with average recovery of 98% with a range of 83-108%

Application

Research Category: 
  • Neuroscience 

Research Sub-Category: 
  • Alzheimer′s Disease
  • Neurochemistry
  • Neurotrophins
  • Signaling Neuroscience

The sensitivity of the Single Molecule Counting (SMC®) technology gives the SMC® Human Brain-Derived Neurotrophic Factor (BDNF) High Sensitivity Immunoassay Kit unparalleled performance in the quantification of BDNF in human EDTA plasma, serum, and cerebrospinal fluid.

Features and Benefits

This new kit includes the following features:

  • Improved options for sample prep and incubation
  • Lyophilization of the standard provides the kit at one storage condition
  • Removal of a plate transfer step
  • Increased reagent volumes to accommodate automation
  • Validated to run on both SMCxPRO® and Erenna

Components

  • Assay Buffer
  • BDNF Coated Beads
  • Standard Diluent
  • BDNF Detection Antibody
  • BDNF Standard
  • 10X Wash Buffer
  • Buffer D
  • Elution Buffer B

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Neuroscience biomarker research expands with high-sensitivity immunoassays for low-abundant biomarkers.

Explore high-sensitivity SMC® neurology immunoassays that precisely measure low levels of neurology protein analytes, such as NF-L or Tau proteins.

Explore ultrasensitive immunoassay technology with SMC® technology, including its biomarker detection platform, advantages of high sensitivity assays, and more.

Find immunoassays to explore neuroimmunology and neuroinflammation cytokines and neurodegenerative disease biomarkers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service