Skip to Content
Merck
All Photos(1)

Key Documents

MAT379362152

Matrix Scientific

2-Chloro-5-(4-chlorophenyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
224.09
MDL number:

mp

104-108

Iupac Name

2-chloro-5-(4-chlorophenyl)pyridine

Inchi Code

1S/C11H7Cl2N/c12-10-4-1-8(2-5-10)9-3-6-11(13)14-7-9/h1-7H

InChI key

AHHKONLQEWVLGR-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Product Link

https://www.matrixscientific.com/086305.html

Melting Point

104-108

Pictograms

MSDS

https://www.matrixscientific.com/086305.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library