Skip to Content
Merck
All Photos(1)

Key Documents

MAN509871545

Manchester Organics

Tetrahydrofuran-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
97.12
MDL number:

Pricing and availability is not currently available.

Iupac Name

tetrahydro-3-furancarbonitrile

Inchi Code

1S/C5H7NO/c6-3-5-1-2-7-4-5/h5H,1-2,4H2

InChI key

OANDNNNKGULGJK-UHFFFAOYSA-N

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/E52330

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library