KEY465196979
Key Organics / BIONET
ethyl N-[(Z)-cyano[2-(3-methoxyphenyl)hydrazin-1-ylidene]carbonyl]carbamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
ethyl (2Z)-2-cyano-2-[(3-methoxyphenyl)hydrazono]ethanoylcarbamate
Inchi Code
1S/C13H14N4O4/c1-3-21-13(19)15-12(18)11(8-14)17-16-9-5-4-6-10(7-9)20-2/h4-7,16H,3H2,1-2H3,(H,15,18,19)/b17-11-
InChI key
FXHSFULFXBFMJI-BOPFTXTBSA-N
Purity
90%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/5K-905
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/5K-905
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.