Skip to Content
Merck
All Photos(1)

Key Documents

ENAH659198AD

Enamine

1-(bromomethyl)-2,2-difluoro-3-phenylbicyclo[1.1.1]pentane

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
273.12
MDL number:

Iupac Name

1-(bromomethyl)-2,2-difluoro-3-phenylbicyclo[1.1.1]pentane

Inchi Code

1S/C12H11BrF2/c13-8-10-6-11(7-10,12(10,14)15)9-4-2-1-3-5-9/h1-5H,6-8H2

InChI key

CTZZLRXRSJKAJP-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-7447573

MSDS

https://www.enaminestore.com/catalog/EN300-7447573


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library