Skip to Content
Merck
All Photos(1)

Documents

ENAH049F3207

Enamine

5-azaspiro[2.5]octan-8-ol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
163.65
MDL number:

Iupac Name

5-azaspiro[2.5]octan-8-ol hydrochloride

Inchi Code

1S/C7H13NO.ClH/c9-6-1-4-8-5-7(6)2-3-7;/h6,8-9H,1-5H2;1H

InChI key

HMAMKIIOLHVQFB-UHFFFAOYSA-N

storage temp.

RT

Salt Data

Cl

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-1590221

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-1590221


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service