ENA769508057
Enamine
dispiro[2.0.2^{4}.1^{3}]heptane-7-carbaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
47-49
Iupac Name
dispiro[2.0.2(4).1(3)]heptane-7-carbaldehyde
Inchi Code
1S/C8H10O/c9-5-6-7(1-2-7)8(6)3-4-8/h5-6H,1-4H2
InChI key
UEKVKRRJOCIUOX-UHFFFAOYSA-N
storage temp.
4
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-366112
Melting Point
47-49
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-366112
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.