Skip to Content
Merck
All Photos(1)

Key Documents

ENA295068845

Enamine

5-methyl-5-(thiophen-3-yl)imidazolidine-2,4-dione

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
196.23
MDL number:

mp

152-153

Iupac Name

5-methyl-5-(3-thienyl)-2,4-imidazolidinedione

Inchi Code

1S/C8H8N2O2S/c1-8(5-2-3-13-4-5)6(11)9-7(12)10-8/h2-4H,1H3,(H2,9,10,11,12)

InChI key

PUTWZZJHSOQALV-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-62489

Melting Point

152-153

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://www.enaminestore.com/catalog/EN300-62489


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library