Skip to Content
Merck
All Photos(1)

Key Documents

CIAH999A8B7F

ChemScene LLC (Preferred Partner)

4,6-Dimethylheptan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
144.26
MDL number:

bp

194.5 C at 760 mmHg

mp

6.15 C

Iupac Name

4,6-dimethyl-2-heptanol

Inchi Code

1S/C9H20O/c1-7(2)5-8(3)6-9(4)10/h7-10H,5-6H2,1-4H3

InChI key

YYUGBYFBCFRGNZ-UHFFFAOYSA-N

storage temp.

4C

Purity

95%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/51079-52-8.html

Boiling point

194.5 C at 760 mmHg

Melting Point

6.15 C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0232992.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library