Skip to Content
Merck
All Photos(1)

Key Documents

Z-013

Supelco

Zopiclone-N-oxide solution

100 μg/mL (Methanol with 1% 1 M HCl), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€124.00

€124.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€124.00

About This Item

Empirical Formula (Hill Notation):
C17H17ClN6O4
CAS Number:
Molecular Weight:
404.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€124.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (Methanol with 1% 1 M HCl)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C1N(C2=CC=C(Cl)C=N2)C(OC(N3CC[N+]([O-])(C)CC3)=O)C4=C1N=CC=N4

InChI

1S/C17H17ClN6O4/c1-24(27)8-6-22(7-9-24)17(26)28-16-14-13(19-4-5-20-14)15(25)23(16)12-3-2-11(18)10-21-12/h2-5,10,16H,6-9H2,1H3

InChI key

IPTIKKTXLHVRKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zopiclone N-oxide is a primary, active metabolite of zopiclone in urine as well as a major pharmaceutical impurity of zopiclone known as eszopiclone impurity A. The parent drug, zopiclone, is a non-benzodiazepine hypnotic sold under the trade names Imovane®and ZImovane®and is used in the treatment of insomnia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Imovane is a registered trademark of Sanofi-Aventis
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service