Skip to Content
Merck
All Photos(1)

Key Documents

V-060

Supelco

Coenzyme Q10 solution

1 mg/mL in ethanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€142.00

€142.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€142.00

About This Item

Empirical Formula (Hill Notation):
C59H90O4
CAS Number:
Molecular Weight:
863.34
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€142.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in ethanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

O(C)C1=C(C(=O)C(=C(C1=O)C\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CC\C=C(\CCC=C(C)C)/C)/C)/C)/C)/C)/C)/C)/C)/C)C)OC

InChI

1S/C59H90O4/c1-44(2)24-15-25-45(3)26-16-27-46(4)28-17-29-47(5)30-18-31-48(6)32-19-33-49(7)34-20-35-50(8)36-21-37-51(9)38-22-39-52(10)40-23-41-53(11)42-43-55-54(12)56(60)58(62-13)59(63-14)57(55)61/h24,26,28,30,32,34,36,38,40,42H,15-23,25,27,29,31,33,35,37,39,41,43H2,1-14H3/b45-26+,46-28+,47-30+,48-32+,49-34+,50-36+,51-38+,52-40+,53-42+

InChI key

ACTIUHUUMQJHFO-UPTCCGCDSA-N

General description

Coenzyme Q10, also known as ubiquinone, ubidecarenone, coenzyme Q, CoQ10, CoQ or Q10, is a fat soluble cofactor, which resembles a vitamin and is important to human health as an antioxidant.

Application

Coenzyme Q10 (CoQ10) may be used as a certified reference material (CRM) for the determination of the analyte in biological samples[1][2], pharmaceuticals[3] and vegetable oils[4] by chromatography techniques.
This Certified Spiking Solution® is suitable for use in a variety of testing applications from diagnostic testing of Coenzyme Q10 deficiency to quality control testing of Coenzyme Q10-based dietary supplements or pharmaceutical preparations by HPLC or LC-MS/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of coenzyme Q10 status in blood mononuclear cells, skeletal muscle, and plasma by HPLC with di-propoxy-coenzyme Q10 as an internal standard
Duncan AJ, et al.
Clinical Chemistry, 51(12), 2380-2382 (2005)
Application of derivative spectrophotometry for determination of coenzyme Q10 in pharmaceuticals and plasma
Karpinska J, et al.
Journal of Pharmaceutical and Biomedical Analysis, 17(8), 1345-1350 (1998)
Improved high-performance liquid chromatographic method for the determination of coenzyme Q10 in plasma
Grossi G, et al.
Journal of Chromatography A, 593(1-2), 217-226 (1992)
Determination of coenzyme Q10 and Q9 in vegetable oils
Rodri?guez-Acun?a R, et al.
Journal of Agricultural and Food Chemistry, 56(15), 6241-6245 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service