Skip to Content
Merck
All Photos(1)

Key Documents

T-150

Supelco

Topiramate-D12 solution

1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€3,200.00

€3,200.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€3,200.00

About This Item

Empirical Formula (Hill Notation):
C12D12H9NO8S
CAS Number:
Molecular Weight:
351.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€3,200.00


Estimated to ship on24 May 2025


Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])C1(O[C@H]2CO[C@@]3(COS(N)(=O)=O)OC(O[C@H]3[C@H]2O1)(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C12H21NO8S/c1-10(2)18-7-5-16-12(6-17-22(13,14)15)9(8(7)19-10)20-11(3,4)21-12/h7-9H,5-6H2,1-4H3,(H2,13,14,15)/t7?,8?,9-,12-/m0/s1/i1D3,2D3,3D3,4D3

InChI key

KJADKKWYZYXHBB-FTZSVORRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Suitable uses for this certified solution standard include as an internal standard for quantitation of topiramate levels in biological matrices such as urine or isotope dilution methods by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications. Topiramate, sold under trade name Topamax®, is an antiepileptic drug used for the treatment of seizures. Topiramate in combination with the stimulant phentermine constitute the active compounds in the new weight lose drug Qsymia®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Qsymia is a registered trademark of Vivus, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Topamax is a registered trademark of Johnson & Johnson

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ladan Espandar et al.
Archives of ophthalmology (Chicago, Ill. : 1960), 130(2), 202-208 (2012-02-15)
To evaluate the ability of human adipose-derived stem cells (h-ASCs) to survive and differentiate in corneal stroma. Our experiment consisted of 2 phases. First, we cultured h-ASCs in different types of hyaluronic acid (HA)-derived synthetic extracellular matrixes (sECMs) to determine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service