Skip to Content
Merck
All Photos(1)

Key Documents

T-145

Supelco

delta9-Tetrahydrocannabinolic acid A-D3 (THCA-A-D3) solution

100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€218.00

€218.00


Estimated to ship on11 April 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€218.00

About This Item

Empirical Formula (Hill Notation):
C22H27D3O4
Molecular Weight:
361.49
UNSPSC Code:
41116107
NACRES:
NA.24

€218.00


Estimated to ship on11 April 2025


Request a Bulk Order

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing

format

single component solution

storage temp.

−70°C

Application


  • Regioselective synthesis and application in cannabinoid profiling: The study presents a method for synthesizing delta9-Tetrahydrocannabinolic acid A-D3 (THCA-A-D3) via the reaction of delta9-Tetrahydrocannabinol-D3 with magnesium methyl carbonate, providing a high-purity THCA-A-D3 reference material for analytical research in cannabinoid quantification, crucial for forensic and pharmaceutical applications (Roth et al., 2012).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service